Skip to Content
Merck
All Photos(1)

Documents

MTCB12009

MTC Bio Western Blot Box

L × W × D 3 3/4 in. (9.5 cm) × 1 3/16 in. (3 cm) × 5/8 in. (1.6 cm), clear

Synonym(s):

multi blot box, stacking western blot box

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

clear
polystyrene

packaging

pack of 5 ea

manufacturer/tradename

MTC Bio B1200-9

L × W × D

3 3/4 in. (9.5 cm) × 1 3/16 in. (3 cm) × 5/8 in. (1.6 cm)

Looking for similar products? Visit Product Comparison Guide

General description

Blot Box, 3 3/4 x 1 3/16 x 5/8in. (9.5 x 3 x 1.6cm), 5/pk
These Blot boxes are correctly matched to common blot sizes, enabling more blots to fit on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. All corners are formed at 90˚ angles and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Black Blot boxes are light-blocking opaque for use with light sensitive samples.

Application

Protean 3 (cut up)

Features and Benefits

  • A perfect box for every blot
  • Reduce antibody usage
  • Hinged lid minimizes evaporation
  • For halves, wholes, strips or minis

Legal Information

MTC is a trademark of MTC Bio

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service