MAK178
Enolase Activity Assay Kit
Sufficient for 100 Colorimetric or Fluorometric tests
Synonym(s):
Enolase Detection Kit
About This Item
detection method
colorimetric
fluorometric
relevant disease(s)
immunological diseases; cardiovascular diseases
storage temp.
−20°C
Gene Information
human ... EBI4(387712) , ENO1(2023) , ENO2(2026)
mouse ... EBI4(226265) , ENO1(13806) , ENO2(13807)
rat ... EBI4(292138) , ENO1(24333) , ENO2(24334)
Related Categories
General description
Application
Suitability
Principle
Kit Components Only
- Enolase Assay Buffer
- Peroxidase Substrate, in DMSO
- Enolase Substrate Mix
- Enolase Converter
- Enolase Developer
- Enolase Positive Control
- Hydrogen Peroxide Standard, 0.88 M
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service