Skip to Content
Merck
All Photos(8)

Key Documents

HPA043317

Sigma-Aldrich

Anti-RHCG antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C15orf6, Anti-PDRC2, Anti-RHGK, Anti-Rh family, C glycoprotein, Anti-SLC42A3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:1000-1:2500

immunogen sequence

RYDFEADAHWWSERTHKNLSDMENEFYYRYP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RHCG(51458)

General description

The Rh family C glycoprotein (RHCG) gene is mapped to human chromosome 15q26.1. RHCG is a non-erythrocytic protein and is a member of ammonia transporters (Amt)/methylammonium permeases (MEP)/rhesus (Rh) family of proteins. RHCG is mainly expressed in the connecting tubule, in the collecting ducts of the mammalian nephrons as well as in extra renal tissues.

Immunogen

Rh family, C glycoprotein recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Rh family C glycoprotein (RHCG) facilitates the transport of ammonium across plasma membranes, to inhibit toxic accumulation of ammonium formed particularly during glutamine metabolism. In addition, it also increases acid secretion in the collecting duct (CD) by stimulating the transport of not only NH3 but also CO2 across the membranes of CD cells. Loss of gene expression leads to the development of human oesophageal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST81969

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weighted gene co-expression based biomarker discovery for psoriasis detection
Sundarrajan S, et al.
Gene, 593, 225?234-225?234 (2016)
RhCG is downregulated in oesophageal squamous cell carcinomas, but expressed in multiple squamous epithelia
Chen BS, et al.
European Journal of Cancer, 38(14), 1927-1936 (2002)
Functional reconstitution into liposomes of purified human RhCG ammonia channel
Mouro-Chanteloup I, et al.
PLoS ONE, 5(1), e8921-e8921 (2010)
Mechanism of NH4+ recruitment and NH3 transport in Rh proteins
Baday S, et al.
Structure, 23(8), 1550-1557 (2015)
Relative CO 2/NH 3 permeabilities of human RhAG, RhBG and RhCG
Geyer RR, et al.
The Journal of Membrane Biology, 246(12), 915-926 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service