Skip to Content
Merck
All Photos(7)

Key Documents

HPA018198

Sigma-Aldrich

Anti-SIGLEC6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD327 antigen, Anti-CD33 antigen-like 1, Anti-CDw327, Anti-OB-BP1, Anti-Obesity-binding protein 1, Anti-Sialic acid-binding Ig-like lectin 6 precursor, Anti-Siglec-6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:1000- 1:2500

immunogen sequence

LPTTLPASYYGYGYWFLEGADVPVATNDPDEEVQEETRGRFHLLWDPRRKNCSLSIRDARRRDNAAYFFRLKSKWMKYGYTSSKLSVRVMALTHRPNISIPGTLESGHPSNLTCSVPWVCEQGT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIGLEC6(946)

Looking for similar products? Visit Product Comparison Guide

General description

The gene sialic acid-binding Ig-like lectin-6 (SIGLEC6) is mapped to human chromosome 19q13.3. SIGLEC6 is a trans-membrane receptor in the CD33-like family of siglecs that binds sialyl-TN glycans and leptin. In humans, the protein is expressed in the placenta (particularly trophoblasts) and circulating human B cells.

Immunogen

Sialic acid-binding Ig-like lectin 6 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Glycodelin-A interaction with sialic acid-binding Ig-like lectin-6 (SIGLEC6) inhibits trophoblast invasiveness by down-regulating extracellular signal-regulated kinases/c-Jun signalling pathway. SIGLEC6 along with leptin is involved in excess proliferation and invasion of gestational trophoblastic disease. SIGLEC6 is up-regulated in preeclampsia, which is one of the main reasons of maternal and perinatal morbidity and mortality and is results in abnormalities in placenta.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71739

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lixin Li et al.
Cellular & molecular immunology, 1(6), 408-415 (2005-11-19)
Modulation by balancing activating and inhibitory receptors constitutes an important mechanism for regulating immune responses. Cells that are activated following ligation of receptors bearing immunoreceptor tyrosine-based activation motifs (ITAMs) can be negatively regulated by other receptors bearing immuno-receptor tyrosine-based inhibition
Kristen K Rumer et al.
Endocrine-related cancer, 19(6), 827-840 (2012-10-24)
Sialic acid immunoglobulin-like lectin (Siglec)-6 is a transmembrane receptor that binds leptin. Leptin is an obesity-associated peptide hormone overexpressed in gestational trophoblastic disease (GTD). GTD encompasses several placental abnormalities that range from benign to malignant. Among GTD, molar placentas are
Kristen K Rumer et al.
Reproductive sciences (Thousand Oaks, Calif.), 20(6), 646-653 (2012-11-23)
Sialic acid immunoglobulin-like lectin (Siglec)-6 is a transmembrane receptor that binds sialyl-TN glycans and leptin. Among eutherian mammals, only human placentas express Siglec-6. Previous work has implicated Siglec-6 in preeclampsia (PE). Preeclampsia, a leading cause of maternal and perinatal morbidity
Els C M Brinkman-Van der Linden et al.
Glycobiology, 17(9), 922-931 (2007-06-21)
CD33-related-Siglecs are lectins on immune cells that recognize sialic acids via extracellular domains, and deliver negative signals via cytosolic tyrosine-based regulatory motifs. We report that while Siglec-6/OB-BP1 (which can also bind leptin) is expressed on immune cells of both humans
Kevin K W Lam et al.
The Journal of biological chemistry, 286(43), 37118-37127 (2011-09-02)
During placentation, the cytotrophoblast differentiates into the villous cytotrophoblast and the extravillous cytotrophoblast. The latter invades the decidualized endometrium. Glycodelin-A (GdA) is abundantly synthesized by the decidua but not the trophoblast. Previous data indicate that GdA suppresses the invasion of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service