Skip to Content
Merck
All Photos(3)

Key Documents

HPA010657

Sigma-Aldrich

Anti-LMTK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AATYK2, Anti-BREK, Anti-KIAA1079, Anti-KPI-2, Anti-KPI2, Anti-LMR2, Anti-PPP1R100, Anti-cprk

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

MELNGVQADFKPATLSSSLDNPKESVITGHFEKEKPRKIFDSEPLCLSDNLMHQDNFDPLNVQELSENFLFLQEKNLLKGSLSSKEHINDLQTELKNAGFTEAMLETSCRNSLDTELQFAENKPGLSLLQENV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMTK2(22853)

General description

LMTK2 (lemur tyrosine kinase 2) contains two transmembrane domains at its N-terminus, a kinase domain and a very long C-terminal tail domain with serine/threonine/tyrosine kinase activity. The gene is mapped to human chromosome 7q21.3.

Immunogen

Serine/threonine-protein kinase LMTK2 precursor recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

LMTK2 (lemur tyrosine kinase 2) gene encodes a member of the protein tyrosine kinase family. The encoded transmembrane protein binds to the WWY site in the C-terminal tail of myosin-VI. This LMTK2–myosin-VI complex plays a role in endocytic and exocytic membrane trafficking pathways. The complex also regulates tubule formation. The encoded protein also serves a binding partner of the p35-activator subunit for cyclin-dependent kinase 5, of protein phosphatase 1C and its small inhibitor protein 2.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71447

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isha Dey et al.
Biochemistry and biophysics reports, 12, 140-150 (2017-11-02)
Regulatory elements present in the promoter of a gene drive the expression of the gene in response to various stimuli. Lemur Tyrosine Kinase 2 (LMTK2) is a membrane-anchored Serine/Threonine kinase involved in endosomal protein trafficking and androgen signaling amongst other
Margarita V Chibalina et al.
Journal of cell science, 120(Pt 24), 4278-4288 (2007-11-22)
Myosin VI is an actin-based retrograde motor protein that plays a crucial role in both endocytic and secretory membrane trafficking pathways. Myosin VI's targeting to and function in these intracellular pathways is mediated by a number of specific binding partners.
Kalpit Shah et al.
Oncotarget, 6(16), 14233-14246 (2015-05-27)
Progression from early forms of prostate cancer to castration-resistant disease is associated with an increase in signal transduction activity. The majority of castration-resistance cancers persist in the expression of the androgen receptor (AR), as well as androgen-dependent genes. The AR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service