Skip to Content
Merck
All Photos(4)

Key Documents

HCLHPVDR

Sigma-Aldrich

HPV/p16 Analyte ControlDR

Cell line controls for immunohistochemistry and in situ hybridization

Synonym(s):

HPV/p16 Dynamic Range Analyte Control

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.47

biological source

human cell culture

packaging

pkg of 1 ea cell microarray block (HCL003)
pkg of 2 slides (HCL001)
pkg of 5 slides (HCL002)

manufacturer/tradename

Histocyte

storage temp.

2-8°C

General description

The HPV/p16 Analyte ControlDR has four cell line cores providing a full range of expression for Human Papilloma Virus (HPV): high, medium, low, and negative. The same cell lines also demonstrate high homogenous, high heterogenous and negative expression of p16.
HistoCyte Laboratories control slides and blocks are made from cultured human cell lines to provide the required range of expression of a specific biomarker. The cell lines are processed using proprietary methods that allow for the creation of a high density embedded cell block with cores that retain their cellular morphology. Available as pre-cut unstained slides or as cell microarray blocks for sectioning.

Application

Ideal for use as a same slide control for HPV in situ hybridization (ISH) and p16 immunohistochemistry (IHC).
The HHPV/p16 Analyte ControlDR histology slides have four cell line cores providing a dynamic range of expression for HPV type 16 and 18: high, medium, low and negative. The same cell lines also demonstrate high homogenous, high heterogenous and negative expression of p16. Analyte controls are placed at one end of slide, allowing sample application on same slide for valid comparison.
The HPV/p16 Analyte ControlDR cell microarray block contains four cell line cores that are 2 mm in diameter and between 3.0-3.5 mm in length. The cell microarray block may yield over 300 sections depending on thickness.

Preparation Note

Cell microarray blocks are fixed in 10% neutral buffered formalin and embedded in paraffin wax. Pre-cut unstained slides are prepared from 4 μm thick sections, mounted on positively charged slides, and dried at 37 °C overnight.

Other Notes

For certificates of analysis, contact info@histocyte.com
HPV/p16 Analyte ControlDR brochure is available from HistoCyte Laboratories

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service