Skip to Content
Merck
All Photos(1)

Key Documents

EP951020460

Eppendorf twin.tec® PCR Plates 96

skirted, size 150 μL, blue, pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
size:
150 μL
96 well
sterility:
non-sterile
feature:
skirt
skirted

sterility

non-sterile

feature

skirt
skirted

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 951020460

technique(s)

PCR: suitable

size

150 μL
96 well

color

blue

Looking for similar products? Visit Product Comparison Guide

General description

Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.
The Eppendorf twin.tec® PCR plate offers a combination of thin-walled polypropylene wells and robust polycarbonate frames. The design allows for optimal heat transfer and high rigidity, respectively. The plates have raised well rims that enable effective sealing, reducing the risk of cross-contamination. They are batch-tested and certified to be free of common polymerase chain reaction (PCR) contaminants. With twin.tec PCR plates, you can have maximum confidence in your results. The plates also provide simplicity and convenience with features like purity, rigidity, and color options for organizing workflows. They are suitable for automation and high-throughput processes. Choose the Eppendorf twin.tec® PCR plates for reliable PCR assays.

Application

Eppendorf twin.tec® PCR Plates 96 is suitable for use in:
  • standard PCR
  • real-time PCR
  • high throughput PCR
  • small volume sample handling
PCR

Features and Benefits

  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
  • The rim of the product is optimized for barcodes and automated work processes.
  • It conforms to the SBS footprint recommendations (127.76 × 85.47 mm ±0.25 mm).
  • Plates are stackable.
  • The skirt with eight holes facilitates plate positioning and removal from the thermal cycler block.
  • The low-profile design enhances PCR efficiency, particularly for small sample volumes.
  • The maximum well volume is 150 µL when used with cap strips.
  • The product is well-suited for quantitative real-time PCR, featuring the OptiTrack® matrix for quicker sample identification and reduced pipetting errors.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service