Skip to Content
Merck
All Photos(3)

Documents

EP6333000022

Eppendorf® Mastercycler® Nexus Thermal Cyclers

Nexus, complete unit, aluminum block, AC/DC input 115 V, 50/60 Hz (US)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102404
UNSPSC Code:
41102421

feature

CSA and CE compliant
Nexus
aluminum block
complete unit
holds 96?×?0.2?mL PCR tubes, 1 PCR plate 8?×?12 or 38?×?0.5 mL PCR tubes
unit can control 2 other units in network

manufacturer/tradename

Eppendorf® 6333000022 (US)

parameter

37-110 °C temp. range (Lid)
4-99 °C temp. range (control range)

technique(s)

PCR: suitable

W × D × H

25 cm × 41.2 cm × 32.1 cm

weight

10.9 kg

Looking for similar products? Visit Product Comparison Guide

General description

Flexlid Concept Due to the flexlid concept, the cycler can deal with all types of PCR consumables, from low volume all the way up to 0.5 mL PCR tubes. Of course, all common PCR plates fit – and if your throughput becomes even higher, you can just connect one or two Mastercycler nexus eco to your Mastercycler nexus
Product Information
With the versatile Mastercycler nexus you can choose the PCR consumables that best suit your assay. The PCR cycler nexus can accommodate 96-well PCR plates, 0.2 mL PCR tubes, 0.2 mL PCR tube strips and 0.5 mL PCR tubes. With a noise level of less than 40 dB[a], the nexus is very quiet and energy-efficient at the same time.

In the age of networking, the new Mastercycler nexus is your reliable companion when it comes to the daily routine of PCR. It is easy to use, does not need much space or energy and sends you an e-mail when it is done – what else do you need from a PCR cycler?

Certified Quality
All Eppendorf cyclers follow these quality guidelines:


  • Individual, documented quality control certificates
  • Calibration accordingly to national and international standards: NIST (USA), DKD/PTB (Germany), UKAS/NPL (UK)
  • UL/cUL listed

The Mastercycler nexus is a whisper quiet PCR machine. Low noise levels make working in your lab more pleasant and enjoyable, particularly when you are thinking about your next big breakthrough findings….

Application

Standard PCR, Fast PCR, PCR optimization

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service