Documents
EP022822101
Rotor Spacer
required for stacking rotors Eppendorf® F-45-72-8 and F-45-48-11 rotors
About This Item
UNSPSC Code:
41103909
feature
required for stacking rotors Eppendorf® F-45-72-8 and F-45-48-11 rotors
manufacturer/tradename
Eppendorf® 022822101
technique(s)
centrifugation: suitable
Related Categories
Legal Information
Eppendorf is a registered trademark of Eppendorf AG