Skip to Content
Merck
All Photos(1)

Key Documents

EHU030131

Sigma-Aldrich

MISSION® esiRNA

targeting human ROCK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATTCTGAGCAACTGGCTCGTTCAATTGCTGAAGAACAATATTCTGATTTGGAAAAAGAGAAGATCATGAAAGAGCTGGAGATCAAAGAGATGATGGCTAGACACAAACAGGAACTTACGGAAAAAGATGCTACAATTGCTTCTCTTGAGGAAACTAATAGGACACTAACTAGTGATGTTGCCAATCTTGCAAATGAGAAAGAAGAATTAAATAACAAATTGAAAGATGTTCAAGAGCAACTGTCAAGATTGAAAGATGAAGAAATAAGCGCAGCAGCTATTAAAGCACAGTTTGAGAAGCAGCTATTAACAGAAAGAACACTCAAAACTCAAGCTGTGAATAAGTTGGCTGAGATCATGAATCGAAAAGAACCTGTCAAGCGTGGTAATGACACAGATGTGCGGAGAAAAGAGAAGGAGAATAGAAAGCTACATATGGAGCTTAAATCTGAACGTGAGAAATTGACCCAGCAGATGATCAAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cecilia Dyberg et al.
Cancers, 12(1) (2020-01-01)
Medulloblastoma is one of the most common malignant brain tumor types in children, with an overall survival of 70%. Mortality is associated with metastatic relapsed tumors. Rho-associated kinases (ROCKs), important for epithelial-mesenchymal transition (EMT) and proper nervous system development, have
Ye Wang et al.
PloS one, 11(1), e0146646-e0146646 (2016-01-09)
Some recent studies suggest that multiple miRNAs might regulate neurogenic transdifferentiation of mesenchymal stromal cells (MSCs). In the present study, we hypothesized that the miR-124 can repress the expression of RhoA upon the neurogenesis of adipose derived MSCs (ADMSCs). MiRNA
Sarah Theresa Boyle et al.
Nature cell biology, 22(7), 882-895 (2020-05-27)
It is well accepted that cancers co-opt the microenvironment for their growth. However, the molecular mechanisms that underlie cancer-microenvironment interactions are still poorly defined. Here, we show that Rho-associated kinase (ROCK) in the mammary tumour epithelium selectively actuates protein-kinase-R-like endoplasmic
Ran You et al.
Clinical science (London, England : 1979), 134(12), 1357-1376 (2020-06-04)
Non-specific inhibition of Rho-associated kinases (ROCKs) alleviated renal fibrosis in the unilateral ureteral obstruction (UUO) model, while genetic deletion of ROCK1 did not affect renal pathology in mice. Thus, whether ROCK2 plays a role in renal tubulointerstitial fibrosis needs to
Ming-Ming Ma et al.
British journal of pharmacology, 173(3), 529-544 (2015-11-13)
Angiotensin II (AngII) induces migration and growth of vascular smooth muscle cell (VSMC), which is responsible for vascular remodelling in some cardiovascular diseases. Ang II also activates a Cl(-) current, but the underlying mechanism is not clear. The A10 cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service