Key Documents
CLS6795610D
Corning® stirrer with digital display
AC/DC input 120 V AC, US 3-pin plug, plate L × W 10 in. × 10 in.
Synonym(s):
magnetic stirrer, magnetic stirrers, stirrers
About This Item
UNSPSC Code:
41103803
NACRES:
NB.73
packaging
case of 1
manufacturer/tradename
Corning 6795-610D
availability
not available in EU
parameter
60-1150 rpm speed
plate L × W
10 in. × 10 in.
General description
  • Digital LED stir speed display indicates when set speed has been reached
  • Microprocessor maintains consistent and repeatable stir speed settings
  • Extremely durable and heat resistant Pyroceram glass-ceramic top
  • Small footprint and low profile
  • Meets UL and cUL standards
Other Notes
individual units are not routinely calibrated, rpm speeds are approximate only
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)