CLS440140
Corning® boss head support clamp
Synonym(s):
lab equipment accessory
All Photos(2)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.76
Recommended Products
material
aluminum
manufacturer/tradename
Corning 440140
L
58 mm
General description
Corning offers a 2 in. long right angle black anodized aluminum Boss Head clamp that will accept up to 7/16 in. diameter support rods. It is designed to be used with the Corning Temperature probe support clamp.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)