CLS3779
Corning® microcarriers
enhanced attachment, weight 10 g
Synonym(s):
cell culture flasks, tissue culture flasks
All Photos(1)
About This Item
feature
enhanced attachment
packaging
pack of 1
manufacturer/tradename
Corning 3779
weight
10 g
General description
Corning Microcarriers are designed to simplify your scale-up processes. The microcarriers are sterile, ready to use, and available in closed systems packaging to be used directly with bioreactors.
Features:
Features:
- USP Class VI polystyrene material provides a consistent platform
- Level (SAL) of 10-6
- Certified nonpyrogenic
The enhanced attachment surface treatment infuses the surface of the microcarriers with oxygen, improving cell attachment, maximizing cell yield and viability. Also available as a closed system solution.
Legal Information
Corning is a registered trademark of Corning, Inc.