PE/CA-PJ34 (clone C12)
97062513, human oral cavity, squamous cell carcinoma. Epithelial morphology.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
product name
PE/CA-PJ34 (clone C12), 97062513
biological source
human mouth
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human basaloid squamous cell carcinoma
Cell Line Description
The human basaloid squamous cell line PE/CA-PJ34 (clone 12) has been established from the oral cavity of a 60 year old male patient in December 1996. The cells, that are also known as Plaepi 34, express cytokeratin.
Application
Study of human squamous carcinoma
Culture Medium
IDMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:2 to 1:8 i.e. seeding a 1-4 x 10,000 cells/cm using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service