Recommended Products
Product Name
RIN-5F, 95090402
biological source
rat pancreas
growth mode
Adherent
karyotype
Not specified
morphology
Not specified
products
Insulin, L-dopa-decarboxylase
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Cell Line Description
Application
Culture Medium
Subculture Routine
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
ECACC pancreatic cancer cell lines for various applications including drug response studies.
ECACC pancreatic cancer cell lines for various applications including drug response studies.
ECACC pancreatic cancer cell lines for various applications including drug response studies.
ECACC pancreatic cancer cell lines for various applications including drug response studies.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service