Skip to Content
Merck
All Photos(1)

Key Documents

P3HR-1

86010701, human lymph, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

P3HR-1, 86010701

biological source

human lymph

packaging

tube of 5 μg 86010701-DNA-5UG
pkg of vial of cells 86010701-1VL

growth mode

Suspension

karyotype

2n = 46, hyperdiploid

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Burkitt′s Lymphoma

Cell Line Description

Clonally-derived sub-line of JIYOYE (Sigma Catalogue number. 88071302) containing high levels of EBV. Has been successfully hybridised with the human lymphoid cell line K562 to form the hybrid PUTKO-1. ACDP class II pathogen and should be handled under laboratory containment level 2. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,11
D13S317: 12
D16S539: 10,11
D5S818: 12
D7S820: 8,10
THO1: 7,9
TPOX: 6,8
vWA: 15,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service