BRL 3A
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 85111503, rat liver, Epithelial
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
rat liver
packaging
tube of 5 μg 85111503-DNA-5UG
pkg of vial of cells 85111503-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial
products
MSA factors
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Buffalo Rat liver
Cell Line Description
Originally isolated by primary cloning. Serum-free medium conditioned by the cells produces a family of polypeptides termed multiplication stimulating activity (MSA).
Application
Growth factor studies
Culture Medium
Coon′s modified Ham′s F12 + 2mM Glutamine + 5% Foetal Bovine Serum (FBS) or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol + 5% Foetal Bovine Serum (FBS) + 2mM Glutamine.
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service