Skip to Content
Merck
All Photos(1)

Key Documents

KARPAS 231

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 06120601, human blood (leukemia), Single round to polymorphic cells growing in suspension

Synonym(s):

KARPAS, 231

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human blood (leukemia)

packaging

tube of 5 μg 06120601-DNA-5UG
pkg of vial of cells 06120601-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Single round to polymorphic cells growing in suspension

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

Cell Line Origin

Human B-cell leukemia

Cell Line Description

Established from peripheral blood of a 71 year-old woman at diagnosis of B-cell acute lymphoblastic leukemia (ALL). No rearrangement of C-MYC DNA sequences.

DNA Profile

Not specified

Culture Medium

RPMI 1640 + 2 mM Glutamine + 10% Fetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 0.5 - 2 x1000,000 cells/ml; 5% CO2; 37 °C. Doubling time approximately 60 hours. Split saturated culture 1:2 every 2-4 days. Cells are single round to polymorphic and grow in suspension.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service