Key Documents
AXYFXF20RS
Pipette filter tips for Beckman® FX Robotics System
racked, capacity 20 μL, pkg of 5x960tips/cs, sterile
Synonym(s):
pipet tips
About This Item
UNSPSC Code:
41121601
NACRES:
NB.02
manufacturer/tradename:
Axygen Inc FXF-20-R-S
Corning
capacity:
20 μL
sterility:
sterile
packaging:
pkg of 5x960tips/cs
sterility
sterile
manufacturer/tradename
Axygen Inc FXF-20-R-S
Corning
packaging
pkg of 5x960tips/cs
capacity
20 μL
General description
  • Axygen® 96-well tips are optimized for use with Beckman Coulter, Biomek FX, Biomek NX, Biomek 3000, and Multimek AP-96, SPAN-8, MP20, MP200, and AP-384 head.
  • Clear polypropylene tips are free of RNase-/DNase-/human DNA
  • Features an aerosol-resistant filter.
  • Automation tips are manufactured to stringent specifications and assembled via automation to ensure consistent performance and quality.
Other Notes
Browse all pipette tips
Legal Information
Axygen is a registered trademark of Corning, Inc.
Beckman is a registered trademark of Beckman Coulter, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)