Recommended Products
description
For In Vitro Diagnostic Use in Select Regions
Quality Level
form
aqueous solution
manufacturer/tradename
Cell Marque®
packaging
kit of (containing 100 mL each of predilute amplifier and polymer detector (962D-30))
kit of (containing 50 mL each of predilute amplifier and polymer detector (962D-20))
kit of (containing 7 mL each of predilute amplifier and polymer detector (962D-10))
technique(s)
immunohistochemistry: suitable
storage temp.
2-8°C
General description
Quality
Canada - IVD
European Union - IVD
Japan - RUO
Physical form
Preparation Note
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1
Storage Class Code
12 - Non Combustible Liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service