Skip to Content
Merck
All Photos(1)

Key Documents

93205

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide phosphate disodium salt

≥97% (calc. based on dry substance, enzymatic)

Synonym(s):

β-NADH phosphate disodium salt, β-NADP-Na2, Triphosphopyridine nucleotide disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N7Na2O17P3
CAS Number:
Molecular Weight:
787.37
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥97% (calc. based on dry substance, enzymatic)

form

powder

impurities

≤7% water

cation traces

Ca: ≤1000 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤20 mg/kg
K: ≤1000 mg/kg
Mg: ≤500 mg/kg
Mn: ≤350 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤50 mg/kg

storage temp.

−20°C

SMILES string

[Na+].[Na+].NC(=O)c1ccc[n+](c1)C2O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](OP(O)([O-])=O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@@H](O)[C@H]2O

InChI

1S/C21H28N7O17P3.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(44-46(33,34)35)14(30)11(43-21)6-41-48(38,39)45-47(36,37)40-5-10-13(29)15(31)20(42-10)27-3-1-2-9(4-27)18(23)32;;/h1-4,7-8,10-11,13-16,20-21,29-31H,5-6H2,(H7-,22,23,24,25,32,33,34,35,36,37,38,39);;/q;2*+1/p-2/t10-,11-,13-,14-,15-,16-,20?,21-;;/m1../s1

InChI key

WSDDJLMGYRLUKR-FHQWUTKFSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Nicotinamide adenine dinucleotide phosphate (NADP) and NADPH form a redox pair. NADP/NADPH is a coenzyme that supports redox reactions via the transport of electrons in a vast array of applications, especially anaerobic reactions such as lipid and nucleic acid synthesis. NADP/NADPH is a coenzyme couple in various cytochrome P450 systems and oxidase/reductase reaction systems, such as the thioredoxin reductase/thioredoxin system.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imad Al Ghouleh et al.
Free radical biology & medicine, 51(7), 1271-1288 (2011-07-05)
Reactive oxygen species (ROS) are involved in numerous physiological and pathophysiological responses. Increasing evidence implicates ROS as signaling molecules involved in the propagation of cellular pathways. The NADPH oxidase (Nox) family of enzymes is a major source of ROS in
Augusto C Montezano et al.
Basic & clinical pharmacology & toxicology, 110(1), 87-94 (2011-09-03)
The healthy endothelium prevents platelet aggregation and leucocyte adhesion, controls permeability to plasma components and maintains vascular integrity. Damage to the endothelium promotes endothelial dysfunction characterized by: altered endothelium-mediated vasodilation, increased vascular reactivity, platelet aggregation, thrombus formation, increased permeability, leucocyte
John E Biaglow et al.
Cancer biology & therapy, 4(1), 6-13 (2005-02-03)
Thioredoxin reductase (TRX) is a selenoprotein that reduces oxidized protein substrates in an NADPH-dependent process (cf. Fig. 1). The thioredoxins (TX) are a family of small redox active proteins that undergo reversible oxidation/reduction and help to maintain the redox state
Sang-Choul Im et al.
Archives of biochemistry and biophysics, 507(1), 144-153 (2010-11-09)
Cytochrome P450 2B4 is a microsomal protein with a multi-step reaction cycle similar to that observed in the majority of other cytochromes P450. The cytochrome P450 2B4-substrate complex is reduced from the ferric to the ferrous form by cytochrome P450
Snigdha Shaw et al.
Scientific reports, 10(1), 3458-3458 (2020-02-28)
The underlying mechanism regulating hypoxia induced alteration in female steroid hormones is first time explored in this study. To understand the mechanistic approach, female Sprague- Dawley rats were exposed to acute and chronic hypobaric hypoxia (282 mm-Hg, ~7620 m, 6 hours, 3 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service