Skip to Content
Merck
All Photos(1)

Key Documents

321R-2

Sigma-Aldrich

PLAP (EP194) Rabbit Monoclonal Primary Antibody

Synonym(s):

Placental Alkaline Phosphatase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
clone:
EP194, monoclonal
application:
IHC (p)
species reactivity:
human
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100 (concentrated)
citations:
7

biological source

rabbit

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP194, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (321R-24)
vial of 0.1 mL concentrate Research Use Only (321R-24-RUO)
vial of 0.5 mL concentrate (321R-25)
vial of 1.0 mL concentrate (321R-26)
vial of 1.0 mL concentrate Research Use Only (321R-26-RUO)
vial of 1.0 mL pre-dilute Research Use Only (321R-27-RUO)
vial of 1.0 mL pre-dilute ready-to-use (321R-27)
vial of 7.0 mL pre-dilute ready-to-use (321R-28)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (321R-28-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:100 (concentrated)

isotype

IgG

control

placenta

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... ALPP(250)

General description

Placental alkaline phosphatase (PLAP) is normally produced by primordial germ cells and syncytiotrophoblasts, and the detection of its expression has been useful in identifying germ cell tumors. Anti-PLAP antibody immunoreacts with germ cell tumors and can discriminate between these and other neoplasms. Somatic neoplasms e.g. breast, gastrointestinal, prostatic and urinary cancers may also immunoreact with antibodies to PLAP. Anti-PLAP positivity in conjunction with keratin negativity favors seminoma over carcinoma. Germ cell tumors are usually keratin positive, but they regularly fail to stain with anti-EMA, whereas most carcinomas stain with anti-EMA. Anti-PLAP has been useful in the diagnosis of gestational trophoblastic disease. Complete hydatidiform mole shows strong expression of hCG and weak expression of PLAP. Weak hCG and strong PLAP expression is found in partial hydatidiform mole. Choriocarcinoma presents strong expression of hCG and weak expression of hPL and PLAP.8 In addition to its role as a germ cell marker, PLAP may also be used as a myogenic marker in identifying soft tissue tumors.

Quality


IVD

IVD

IVD

RUO

Linkage

PLAP Positive Control Slides, Product No. 321S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuping Gao et al.
International journal of clinical and experimental pathology, 7(10), 6965-6972 (2014-11-18)
Primary central nervous system (CNS) germ cell tumors (GCTs) are a rare heterogeneous group of lesions, which the clinicopathological features have a marked degree of heterogeneity comparing with that of gonadal GCTs. Accurately diagnosing CNS GCTs might be extremely difficult
Fenghua Wang et al.
The American journal of surgical pathology, 33(10), 1529-1539 (2009-07-04)
Extragonadal yolk sac tumors (YSTs; primary and metastatic) are rare but are malignant germ cell tumors. Pathologic diagnosis of extragonadal YSTs can be challenging without immunohistochemical markers but markers used for diagnosing these tumors such as placental-like alkaline phosphatase (PLAP)
A Lösch et al.
Acta obstetricia et gynecologica Scandinavica, 75(8), 753-756 (1996-09-01)
The gestational trophoblastic disease summarizes all types of hydatidiform moles, placental site trophoblastic tumor and choriocarcinoma. It is of clinical relevance to distinguish between complete hydatidiform mole and partial hydatidiform mole to predict prognosis of recurrency of molar pregnancy and
Jeffrey D Goldsmith et al.
The American journal of surgical pathology, 26(12), 1627-1633 (2002-12-03)
Placental alkaline phosphatase (PLAP) is normally produced by primordial germ cells and syncytiotrophoblasts, and the detection of its expression has been useful in the diagnosis of germ cell tumors. We have recently observed PLAP immunoreactivity in normal human adult and
M R Wick et al.
Human pathology, 18(9), 946-954 (1987-09-01)
Placental-like alkaline phosphatase (PLAP) activity has been reported in various human neoplasms of both somatic and germ cell types. The expression of PLAP was examined with a polyclonal antibody and the immunoperoxidase technique in formalin-fixed, paraffin-embedded sections of 37 germ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service