Skip to Content
Merck
All Photos(7)

Key Documents

243973

Sigma-Aldrich

Sodium bisulfite

ACS reagent

Synonym(s):

Sodium hydrogensulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaHSO3
CAS Number:
Molecular Weight:
104.06
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals

grade

ACS reagent

Quality Level

form

powder or crystals

concentration

≥58.5% SO2

impurities

≤0.005% insolubles

pH

4.3 (10 g/L)

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].OS([O-])=O

InChI

1S/Na.H2O3S/c;1-4(2)3/h;(H2,1,2,3)/q+1;/p-2

InChI key

DWAQJAXMDSEUJJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium bisulfite is a widely used industrial chemical as a reducing agent, catalyst, and preservative in pharmaceuticals and foods due to its antioxidative and antibacterial effects.
The aqueous solutions of sodium bisulfite are acidic in nature. Degradation of sodium bisulfite with acid forms sulfur dioxide gas.

Application

Sodium bisulfite (NaHSO3) has been used as a reagent to compose the immunoprecipitation (IP) buffer to chemically modify DNA and to synthesize arsenolipids (AsL).
It can also be used as a reagent during the synthesis of 5,6-dihydrouracil-6-sulfonate.
Sodium bisulfite can be used as:
  • A reagent to synthesize aldehyde-bisulfite adducts from aromatic and aliphatic aldehydes.
  • A catalyst in the solvent-free etherification of aromatic/aliphatic alcohols to synthesize symmetric and unsymmetric ethers via intermolecular dehydration.
  • An additive in the copper-catalyzed homocoupling of ketoxime carboxylates to synthesize symmetrical pyrroles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ledgard J.
The Preparatory Manual of Explosives., 87-87 (2007)
Synthesis of two new arsenolipids and their identification in fish.
Arroyo-Abad U, et al.
European Journal of Lipid Science and Technology, 118(3), 445-452 (2015)
Reaction of sodium bisulfite with uracil, cytosine, and their derivatives.
H Hayatsu et al.
Biochemistry, 9(14), 2858-2865 (1970-07-07)
Hamid Mostafavi Abdolmaleky et al.
Methods in molecular biology (Clifton, N.J.), 448, 187-212 (2008-03-29)
Although there is evidence to link schizophrenia (SCZ) and bipolar disorder (BD) to genetic and environmental factors, specific individual or groups of genes/factors causative of the disease have been elusive to the research community. An understanding of the molecular aberrations
Hua-Chien Chen et al.
Journal of chromatography. A, 1230, 123-129 (2012-02-22)
The methylation of the promoter region of DNA is an important regulatory mechanism for the downstream gene expression, and the extent of methylation has been linked to cancer formation. In this study, we report a simple method to screen for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service