Skip to Content
Merck
All Photos(1)

Key Documents

221929

Sigma-Aldrich

Iodic acid

ACS reagent, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HIO3
CAS Number:
Molecular Weight:
175.91
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Assay

≥99.5%

form

crystals or chunks

impurities

≤0.01% insolubles
≤0.1% N compounds

ign. residue

≤0.02%

solubility

water: soluble

density

4.63 g/cm3

anion traces

Cl- and Br-: ≤0.02%
iodide (I-): ≤0.01%
sulfate (SO42-): ≤0.015%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

SMILES string

OI(=O)=O

InChI

1S/HIO3/c2-1(3)4/h(H,2,3,4)

InChI key

ICIWUVCWSCSTAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glass formation and unusual hygroscopic growth of iodic acid solution droplets with relevance for iodine oxide particles in the coastal marine boundary layer.
Murray BJ, et al.
Atmospheric Chemistry and Physics Discussions, 12(3), 7879-7908 (2012)
Manometric carbon determination.
Van Slyke DD and Folch J.
The Journal of Biological Chemistry, 136(2), 509-541 (1940)
Raman studies of iodic acid and sodium iodate.
Durig JR, et al.
The Journal of Physical Chemistry, 69(11), 3886-3892 (1965)
The rate of the reaction between iodic and hydriodic acids.
Dushman S.
The Journal of Physical Chemistry, 8(7), 453-482 (1904)
Allen E Haddrell et al.
The journal of physical chemistry. A, 116(40), 9941-9953 (2012-09-18)
The most used instrument in single particle hygroscopic analysis over the past thirty years has been the electrodynamic balance (EDB). Two general assumptions are made in hygroscopic studies involving the EDB. First, it is assumed that the net charge on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service