Skip to Content
Merck
All Photos(1)

Key Documents

Y0000480

Dibrompropamidine diisetionate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30Br2N4O10S2
CAS Number:
Molecular Weight:
722.42
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dibrompropamidine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

InChI

1S/C17H18Br2N4O2.C2H6O4S/c18-12-8-10(16(20)21)2-4-14(12)24-6-1-7-25-15-5-3-11(17(22)23)9-13(15)19;3-1-2-7(4,5)6/h2-5,8-9H,1,6-7H2,(H3,20,21)(H3,22,23);3H,1-2H2,(H,4,5,6)

InChI key

BZACJASHKPPTKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dibrompropamidine diisetionate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[When the treatment is the problem].
Dag Sollesnes Holsen
Tidsskrift for den Norske laegeforening : tidsskrift for praktisk medicin, ny raekke, 129(4), 307-307 (2009-02-24)
Victoria W Y Wong et al.
Eye & contact lens, 33(6 Pt 1), 329-331 (2007-11-13)
To describe a patient with a good visual outcome after prompt treatment of Acanthamoeba keratitis as a complication of overnight orthokeratology lens wear. Interventional case report. A 9-year-old boy experienced pain, photophobia, and redness in his right eye 3 days
M T Suller et al.
The Journal of hospital infection, 43(4), 281-291 (2000-02-05)
Concern has been growing regarding the potential of antibiotic and disinfectant co-resistance in clinically important bacteria. In this study, the susceptibilities of methicillin-resistant Staphylococcus aureus (MRSA) and methicillin-sensitive Staphylococcus aureus (MSSA) to chlorhexidine (CHX), the quaternary ammonium compounds cetylpyridinium chloride
B Rabanal et al.
Journal of chromatography. B, Biomedical sciences and applications, 738(2), 293-303 (2000-03-16)
A quick, simple and reliable analysis method has been developed in order to determine berenil, phenamidine, diampron and dibromopropamidine by capillary zone electrophoresis in samples of serum and urine. In order to define the operation parameters in CZE, we have
R M Richards et al.
Journal of pharmaceutical sciences, 80(9), 861-867 (1991-09-01)
Modifications of antibacterial activity have been demonstrated using combinations of two antibacterials from trimethoprim, sulfonamides (sulfadiazine, sulfamerazine, and silver sulfadiazine), silver nitrate, and dibromopropamidine isethionate, either formulated in a cream base or dissolved in peptone water. The creams were evaluated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service