Skip to Content
Merck
All Photos(1)

Documents

QC1083

Supelco

Oil and Grease

certified reference material

Synonym(s):

synonym description not exact

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

CofA

current certificate can be downloaded

manufacturer/tradename

RTC QCI-029

concentration

in 1-propanol

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup - Solvent

Flash Point(C)

22 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service