All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C15H20O2
CAS Number:
Molecular Weight:
232.32
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
primary reference standard
product line
phyproof® Reference Substance
Assay
≥90.0% (HPLC)
manufacturer/tradename
PhytoLab
application(s)
food and beverages
format
neat
storage temp.
−20°C
SMILES string
C\C1=C/CC\C(C)=C\[C@H]2OC(=O)C(=C)[C@@H]2CC1
InChI
1S/C15H20O2/c1-10-5-4-6-11(2)9-14-13(8-7-10)12(3)15(16)17-14/h5,9,13-14H,3-4,6-8H2,1-2H3/b10-5+,11-9+/t13-,14+/m0/s1
InChI key
HRYLQFBHBWLLLL-AHNJNIBGSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG
Biochem/physiol Actions
Costunolide is a sesquiterpene lactone originally isolated from plants widely used in many herbal medicines. It has a variety of effects. Costunolide is a potent inducer of apoptosis, a potent anti-cancer agent, has anti-inflammatory, anti-viral, anti-fungal, antimycobacterial activity and has been shown to inhibit telomerase activity.
Legal Information
phyproof is a registered trademark of PhytoLab GmbH & Co. KG
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Melanie S Matos et al.
Nutrients, 12(11) (2020-11-25)
Cichorium intybus L. has recently gained major attention due to large quantities of health-promoting compounds in its roots, such as inulin and sesquiterpene lactones (SLs). Chicory is the main dietary source of SLs, which have underexplored bioactive potential. In this
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service