P6667
Poly(ethylene glycol)
average mol wt 10,000
Synonym(s):
Polyethylene glycol, PEG
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
H(OCH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
form
solid
Quality Level
mol wt
average mol wt 10,000
solubility
water: soluble (100 mg/ml)
SMILES string
C(CO)O
InChI
1S/C2H6O2/c3-1-2-4/h3-4H,1-2H2
InChI key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Mari Palviainen et al.
Cancers, 12(11) (2020-11-12)
Cancer alters cell metabolism. How these changes are manifested in the metabolite cargo of cancer-derived extracellular vesicles (EVs) remains poorly understood. To explore these changes, EVs from prostate, cutaneous T-cell lymphoma (CTCL), colon cancer cell lines, and control EVs from
Yow-Jiun Jeng et al.
Journal of molecular signaling, 4, 2-2 (2009-04-30)
Estradiol (E2) mediates various intracellular signaling cascades from the plasma membrane via several estrogen receptors (ERs). The pituitary is an estrogen-responsive tissue, and we have previously reported that E2 can activate mitogen-activated protein kinases (MAPKs) such as ERK1/2 and JNK1/2/3
Ann L Wozniak et al.
Environmental health perspectives, 113(4), 431-439 (2005-04-07)
Xenoestrogens (XEs) are widespread in our environment and are known to have deleterious effects in animal (and perhaps human) populations. Acting as inappropriate estrogens, XEs are thought to interfere with endogenous estrogens such as estradiol (E2) to disrupt normal estrogenic
Po-Ling Kuo et al.
Journal of cell science, 130(1), 292-302 (2016-08-10)
Studying mechanobiology in three-dimensional (3D) cell cultures better recapitulates cell behaviors in response to various types of mechanical stimuli in vivo Stiffening of the extracellular matrix resulting from cell remodeling potentiates many pathological conditions, including advanced cancers. However, an effective
Idalis Villanueva et al.
Acta biomaterialia, 5(8), 2832-2846 (2009-06-11)
The pericellular matrix (PCM) surrounding chondrocytes is thought to play an important role in transmitting biochemical and biomechanical signals to the cells, which regulates many cellular functions including tissue homeostasis. To better understand chondrocytes interactions with their PCM, three-dimensional poly(ethylene
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service