Skip to Content
Merck
All Photos(1)

Key Documents

H2882

Supelco

Hydrocortisone 21-hemisuccinate

analytical standard

Synonym(s):

11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-hemisuccinate, Cortisol 21-hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H34O8
CAS Number:
Molecular Weight:
462.53
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

−20°C

SMILES string

C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)CCC(O)=O)[C@@]4(C)C[C@H](O)[C@H]23

InChI

1S/C25H34O8/c1-23-9-7-15(26)11-14(23)3-4-16-17-8-10-25(32,24(17,2)12-18(27)22(16)23)19(28)13-33-21(31)6-5-20(29)30/h11,16-18,22,27,32H,3-10,12-13H2,1-2H3,(H,29,30)/t16-,17-,18-,22+,23-,24-,25-/m0/s1

InChI key

VWQWXZAWFPZJDA-CGVGKPPMSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaohong Liu et al.
General and comparative endocrinology, 275, 65-72 (2019-02-16)
The pituitary is a central organ of the neuro-endocrine system in fish that plays critical roles in various physiological processes, including stress response and behavior. Although it is known that pituitary hormones can have a direct or indirect influence stimulating
Mark P Yeager et al.
Critical care medicine, 37(10), 2727-2732 (2009-11-04)
There is continuing controversy regarding the effect of glucocorticoids on a systemic inflammatory process. Based ona model of glucocorticoid action that includes both pro- and anti-inflammatory effects, we used the human experimental endotoxemia model to test the hypothesis that a
Noriko Hikita-Watanabe et al.
Acta oto-laryngologica, 130(1), 79-83 (2009-05-14)
The 'tinnitus-rare' group had a poorer prognosis for hearing than the 'tinnitus-often' group in all sudden sensorineural hearing loss (SSNHL), although the 'shorter duration' group had better prognosis than the 'longer duration' when restricted to SSNHL accompanied by tinnitus. This
Ryoukichi Ikeda et al.
Auris, nasus, larynx, 37(1), 114-116 (2009-05-05)
With the increase in the number of patients undergoing warfarin therapy, reports of complications due to such therapy have become frequent. Although upper airway obstruction secondary to bleeding resulting from warfarin therapy is rare, it is a life-threatening complication because
Setsuko Morinaka
Auris, nasus, larynx, 36(5), 606-608 (2009-03-07)
A 73-year-old man presented to our hospital with a sore throat (left-sided) and hiccups. The patient had mucosal swelling and erosions affecting the left posterior pillar, base of tongue, epiglottis, arytenoid, and aryepiglottic fold. As the laryngeal mucosal edema became

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service