Skip to Content
Merck
All Photos(1)

Documents

BCR679

White cabbage (trace elements)

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

Application

White cabbage (trace elements) has been used as a reference material in the analysis of lead and cadmium in biological reference materials using electrothermal atomic absorption spectrometry (ET-AAS).

Analysis Note

For more information please see:
BCR679

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phosphate
Trace Elements: Antimony (Sb), Arsenic (As), Barium (Ba), Boron (B), Cadmium (Cd), Calcium (Ca), Chromium (Cr), Copper (Cu), Iron (Fe), Magnesium (Mg), Manganese (Mn), Mercury (Hg), Molybdenum (Mo), Nickel (Ni), Strontium (Sr), Thallium (Tl), Zinc (Zn)

Matrix Group: Fruits and Vegetables

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Determination of Cd and Pb in biological reference materials by electrothermal atomic absorption spectrometry: A comparison of three ultrasonic-based sample treatment procedures
Maduro.C, et al.
Talanta, 68, 1156-1161 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service