Skip to Content
Merck
All Photos(1)

Key Documents

85432

Supelco

Silylating mixture II according to Horning

for GC derivatization, LiChropur

Synonym(s):

N,O-Bis(trimethylsilyl)acetamide – chlorotrimethylsilane – 1-(trimethylsilyl)imidazole mixture, BSA+TMCS+TMSI 3:2:3, TMSIm/BSA/TMCS 3/3/2 (v/v/v)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
NA.22

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

storage temp.

2-8°C

InChI

1S/C8H21NOSi2.C6H12N2Si.C3H9ClSi/c1-8(9-11(2,3)4)10-12(5,6)7;1-9(2,3)8-5-4-7-6-8;1-5(2,3)4/h1-7H3;4-6H,1-3H3;1-3H3

InChI key

QSBHIAMPMUWWFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Learn more in the Product Information

Components

N,O-Bis(trimethylsilyl)acetamide : chlorotrimethylsilane : 1-(trimethylsilyl)imidazole = 3:3:2 (v/v/v)

Other Notes

One of the most powerful, general silylating mixture

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Heberle et al.
Silylating Agents, 2nd ed. (1995)
Conversion of steroids to trimethylsilyl derivatives for gas phase analytical studies: reactions of silylating reagents.
E M Chambaz et al.
Analytical biochemistry, 30(1), 7-24 (1969-07-01)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service