Skip to Content
Merck
All Photos(1)

Key Documents

80543

Supelco

Antonia Red Dextran 150

Mw 140000-160000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171501
NACRES:
NA.25

biological source

(Leuconostoc mesenteroides B-512F)

Quality Level

form

powder or crystals

mol wt

Mw 140000-160000

manufacturer/tradename

TdB Consultancy AB

extent of labeling

0.001-0.01 (degree of substitution)

loss

≤10% loss on drying

fluorescence

λex 583 nm; λem 602 nm in borate buffer (pH 9.0)

detection method

fluorometric

storage temp.

room temp

General description

Antonia Red dextrans are purple powders that are readily soluble in water or electrolytesolutions. Antonia Red dextran are insoluble in most organic solvents.

Application

Antonia Red dextranes are primarily used for studying permeability and transport in cells and tissues, trace cell lineage and perform vasculature studies. An added benefit is that measurements of the fluorescence provide quantitative data on the permeability of healthy and diseased tissues. Such studies can be performed in real time by intra-vital fluorescence microscopy. The technique offers high sensitivity and concentrations down to 1μg/ml can be detected in tissue fluids. Antonia Red dextranes is brightly fluorescent using the red (Texas Red®) filter in fluorescence microscopy.

Other Notes

The dextran used is essentially a linear α-(1-6)-linked glucose chain with a low percentage (2-5%) of α-(1-3) branches distributed along the chain.

Legal Information

Antonia Red is a trademark of TdB Consultancy AB
Texas Red is a registered trademark of Life Technologies

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service