Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
80457
Trifluoroacetic acid
eluent additive for LC-MS, LiChropur™, ≥99.0% (GC)
Synonym(s):
TFA
About This Item
Recommended Products
vapor density
3.9 (vs air)
Quality Level
vapor pressure
97.5 mmHg ( 20 °C)
Assay
≥99.0% (GC)
form
liquid
technique(s)
LC/MS: suitable
impurities
≤0.05% water
≤50 mg/kg total sulfur (as SO4)
refractive index
n20/D 1.3 (lit.)
bp
72.4 °C (lit.)
mp
−15.4 °C (lit.)
density
1.489 g/mL at 20 °C (lit.)
anion traces
chloride (Cl-): ≤50 mg/kg
sulfite (SO32-): ≤10 mg/kg
cation traces
Ag: ≤0.05 mg/kg
Al: ≤0.1 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.5 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.1 mg/kg
Co: ≤0.05 mg/kg
Cr: ≤0.1 mg/kg
Cu: ≤0.05 mg/kg
Fe: ≤1.0 mg/kg
K: ≤1.0 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤1.0 mg/kg
Mn: ≤0.1 mg/kg
Mo: ≤0.05 mg/kg
NH4+: ≤10 mg/kg
Na: ≤1.0 mg/kg
Ni: ≤0.1 mg/kg
Pb: ≤0.05 mg/kg
Sr: ≤0.05 mg/kg
Tl: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg
UV absorption
λ: 260 nm Amax: ≤0.9
λ: 270 nm Amax: ≤0.1
λ: 280 nm Amax: ≤0.08
λ: 290 nm Amax: ≤0.07
λ: 300 nm Amax: ≤0.06
λ: 320 nm Amax: ≤0.05
SMILES string
OC(C(F)(F)F)=O
suitability
passes test for LC-MS
InChI
1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key
DTQVDTLACAAQTR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Features:
- Extensive QC testing ensures the highest specifications.
- Optimized to improve ionization and resolution.
- Extremely low levels of inorganic and organic impurities.
- Manufactured specifically for accurate and fast LC-MS.
Application
Packaging
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
>212.0 °F - Pensky-Martens closed cup
Flash Point(C)
> 100 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
The article describes the use of difluoroacetic acid (DFA) as an effective alternative to acid modifiers, formic acid (FA) and trifluoroacetic acid (TFA) in the LC-UV/MS analysis of proteins.
The article describes the use of difluoroacetic acid (DFA) as an effective alternative to acid modifiers, formic acid (FA) and trifluoroacetic acid (TFA) in the LC-UV/MS analysis of proteins.
The article describes the use of difluoroacetic acid (DFA) as an effective alternative to acid modifiers, formic acid (FA) and trifluoroacetic acid (TFA) in the LC-UV/MS analysis of proteins.
The article describes the use of difluoroacetic acid (DFA) as an effective alternative to acid modifiers, formic acid (FA) and trifluoroacetic acid (TFA) in the LC-UV/MS analysis of proteins.
Protocols
Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.
Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.
Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.
Straightforward HPTLC-MS analysis of lactose in dairy products (milk or yoghurt) using only protein crash, centrifugation and dilution as sample preparation.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service