Skip to Content
Merck
All Photos(1)

Key Documents

79856

Supelco

Isoborneol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

212-214 °C (subl.) (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H][C@@]12CC[C@@](C)([C@H](O)C1)C2(C)C

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8-,10+/m1/s1

InChI key

DTGKSKDOIYIVQL-MRTMQBJTSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isoborneol is a monoterpene and a known antioxidant, which is a component of several plant essential oils.

Application

Isoborneol may be used as a reference standard for the determination of isoborneol in plasma samples using headspace, solid-phase dynamic extraction coupled to gas chromatography−tandem mass spectrometry (HS-SPDE-GC−MS/MS).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 1 - Skin Irrit. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

200.1 °F - closed cup

Flash Point(C)

93.4 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of four volatile compounds in rat plasma after oral administration of Shexiang Baoxin Pill (SBP) by HS-SPDE-GC-MS/MS and its application to pharmacokinetic studies.
Chang W, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 963(6), 47-53 (2014)
Protective effect of (?) isoborneol against 6-OHDA-induced apoptosis in SH-SY5Y cells.
L.LT,et al.
Cellular Physiology and Biochemistry, 20(6), 1019-1032 (2007)
Antiviral properties of isoborneol, a potent inhibitor of herpes simplex virus type 1.
Armaka M, et al.
Antiviral research, 43(2), 79-92 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service