Skip to Content
Merck
All Photos(1)

Documents

78827

Supelco

TDCPP-d15

analytical standard

Synonym(s):

Phosphoric acid tris(1,3-dichloro-2-propyl-d5 ester), Tris(1,3-dichloro-2-propyl-d5) phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D15Cl6O4P
CAS Number:
Molecular Weight:
446.00
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

SMILES string

O=P(OC(C([2H])([2H])Cl)([2H])C([2H])([2H])Cl)(OC(C([2H])([2H])Cl)([2H])C([2H])([2H])Cl)OC(C([2H])([2H])Cl)([2H])C([2H])([2H])Cl

InChI

1S/C9H15Cl6O4P/c10-1-7(2-11)17-20(16,18-8(3-12)4-13)19-9(5-14)6-15/h7-9H,1-6H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D,8D,9D

InChI key

ASLWPAWFJZFCKF-BXSQCBKHSA-N

General description

TDCPP-d15 is the deuterated analog of tris(1,3-dichloroisopropyl)phosphate (TDCPP), an organophosphorus flame retardant (OPFR). It is generally used in consumer goods, such as plastics, foams, paints, resins, textiles, electronics, and furniture. OPFRs are high-production volume chemicals that are regarded as potential environmental pollutants.

Application

TDCPP- d15 may be used as an analytical reference standard for the determination of the analyte in whole parts of fish and waste-water by various gas-chromatography based techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Carc. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of human metabolites of chlorinated phosphorous flame retardants in wastewater by N-tert-butyldimethylsilyl-N-methyltrifluoroacetamide-derivatization and gas chromatography-high resolution mass spectrometry
Castro V, et al.
Journal of Chromatography A, 1602, 450-457 (2019)
Organophosphorus flame-retardant and plasticizer analysis, including recommendations from the first worldwide interlaboratory study
Brandsma SH, et al.
Trends in Analytical Chemistry, 43, 217-228 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service