Skip to Content
Merck
All Photos(2)

Key Documents

46682

Supelco

Promethazine hydrochloride

VETRANAL®, analytical standard

Synonym(s):

10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2S · HCl
CAS Number:
Molecular Weight:
320.88
Beilstein:
4166397
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

SMILES string

Cl[H].CC(CN1c2ccccc2Sc3ccccc13)N(C)C

InChI

1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H

InChI key

XXPDBLUZJRXNNZ-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Mario Schietroma et al.
Journal of the American College of Surgeons, 220(5), 921-933 (2015-04-05)
This article has been retracted: please see Elsevier Policy on Article Withdrawal (https://www.elsevier.com/about/our-business/policies/article-withdrawal). This article has been retracted at the request of the Editor-in-Chief. An independent statistical analysis based on the summary data tables and statistical results reported in the
Shigeru Hishinuma et al.
Biochemical pharmacology, 91(2), 231-241 (2014-07-30)
Differential binding sites for first- and second-generation antihistamines were indicated on the basis of the crystal structure of human histamine H1 receptors. In this study, we evaluated differences between the thermodynamic driving forces of first- and second-generation antihistamines for human
Cheryl C Y Loh et al.
PloS one, 9(10), e110800-e110800 (2014-10-25)
Chloroquine was a cheap, extremely effective drug against Plasmodium falciparum until resistance arose. One approach to reversing resistance is the inhibition of chloroquine efflux from its site of action, the parasite digestive vacuole. Chloroquine accumulation studies have traditionally relied on
Jung Yeon Kim et al.
Biomedical chromatography : BMC, 29(3), 465-474 (2014-08-01)
The purpose of this study was to develop and validate an ultra-performance liquid chromatography method for simultaneous analysis of 20 antihistamines (illegal additives) in dietary supplements. The limits of detection and quantitation of the method ranged from 1.5 to 2.5
Andrew Curtright et al.
PloS one, 10(1), e0116766-e0116766 (2015-01-15)
Acute and chronic pain conditions are often debilitating, inflicting severe physiological, emotional and economic costs and affect a large percentage of the global population. However, the development of therapeutic analgesic agents based primarily on targeted drug development has been largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service