Skip to Content
Merck
All Photos(1)

Documents

43677

Supelco

Tetrachlorvinphos

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Stirofox

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9Cl4O4P
CAS Number:
Molecular Weight:
365.96
Beilstein:
1890909
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

form

solid

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

97-98 °C (lit.)

storage temp.

−20°C

SMILES string

COP(=O)(OC)O\C(=C/Cl)c1cc(Cl)c(Cl)cc1Cl

InChI

1S/C10H9Cl4O4P/c1-16-19(15,17-2)18-10(5-11)6-3-8(13)9(14)4-7(6)12/h3-5H,1-2H3/b10-5-

InChI key

UBCKGWBNUIFUST-YHYXMXQVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com

Tetrachlorvinphos is a non-systemic insecticide and acaricide that belongs to the group of organophosphates. It is an acetylcholinesterase (AChE) inhibitor causing hyperexcitation and impeding the functioning of the nervous system. Tetrachlorvinphos is used for protection against Diptera in fruit and Lepidoptera in fruit, cotton, maize, rice, tobacco, and vegetables; control of Muscidae in dairies and livestock barns; control of ectoparasites in poultry; control of insects in stored products and insects that damage forests and pastures.
According to the Commission Implementing Regulation (EU) No 2076/2002 of 20th November 2002, tetrachlorvinphos is not approved for use in the European Union (EU) as a plant protection agent, following the Regulation (EC) No. 1107/2009.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Tetrachlorvinphos CRM may also be used as given below:

  • Trace analysis of 160 pesticides in 50 honeybee wax combs samples by gas chromatography combined with triple quadrupole mass spectrometry for quantitation, following a QuEChERS-based method for extraction
  • Multi-residue analysis of 213 pesticides in edible oil samples using an analytical approach combining QuEChERS methodology with gas chromatography-tandem mass spectrometry (GC-MS/MS)
  • Detection of 379 pesticides in human serum samples extracted by QuEChERS method― using liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Multi-residue analysis of 327 pesticides and 17 veterinary drug residues in eight animal feed samples using liquid- and gas- chromatography combined with tandem mass spectrometry (LC-MS/MS and GC-MS/MS)
  • Development of a semi-automated method for the analysis of 261 pesticides and 41 environmental contaminants in catfish muscle samples by QuEChERS sample extraction and low-pressure GC-MS/MS and UHPLC-MS/MS, following an automated robotic micro-SPE clean-up
  • Method development and validation for the determination of 360 pesticide residues in brown rice, orange, spinach, and potato samples by a modified QuEChERS protocol combined with gas chromatography-tandem mass spectrometry (GC-MS/MS)

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service