Skip to Content
Merck
All Photos(1)

Key Documents

24-6770

Sigma-Aldrich

Pyrophosphoric acid

SAJ first grade, ≥90.0%

Synonym(s):

Diphosphoric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2P(O)OP(O)(OH)2
CAS Number:
Molecular Weight:
177.98
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

SAJ first grade

Assay

≥90.0%

form

solid

availability

available only in Japan

technique(s)

ion chromatography: suitable

SMILES string

OP(O)(=O)OP(O)(O)=O

InChI

1S/H4O7P2/c1-8(2,3)7-9(4,5)6/h(H2,1,2,3)(H2,4,5,6)

InChI key

XPPKVPWEQAFLFU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aparajita Choudhury et al.
FEBS letters, 587(6), 590-599 (2013-02-07)
Acanthamoeba polyphaga mimivirus tyrosyl-tRNA synthetase (TyrRSapm) was the first reported aminoacyl-tRNA synthetase of viral origin. The previous crystal structure of TyrRSapm showed a non-canonical orientation of the dimer conformation and the CP1 domain, responsible for dimer formation, displays a major
Vijay Luxami et al.
Dalton transactions (Cambridge, England : 2003), 42(11), 3783-3786 (2013-01-26)
Highly selective detection of pyrophosphate in the presence of inorganic phosphates, halides, acetate, ATP, other nucleotides in neutral aqueous solutions is achieved by a fluorescent Zn-ensemble based system. The implementation of the first molecule based JK-latch sequential logic function is
Florence Mauger et al.
Human mutation, 34(1), 266-273 (2012-11-08)
Here, we describe a high-throughput, single-tube, allele-specific ribonucleotide analog pyrophosphorolysis-activated polymerization (ribo-PAP) PCR multiplex genotyping and resequencing method. An RNA/DNA chimeric PCR product is generated using genomic DNA as starting template, a panel of allele-selective 5'-tagged primers, a reverse primer
Matthew L Fowler et al.
Eukaryotic cell, 11(10), 1249-1256 (2012-08-21)
Acetate kinase (ACK) catalyzes the reversible synthesis of acetyl phosphate by transfer of the γ-phosphate of ATP to acetate. Here we report the first biochemical and kinetic characterization of a eukaryotic ACK, that from the protist Entamoeba histolytica. Our characterization
Xin Su et al.
Chemical communications (Cambridge, England), 49(8), 798-800 (2012-12-12)
A kinetic method has been developed for highly sensitive and selective detection of pyrophosphate anions (PPi) by using a fluorophore labelled single-stranded-DNA-Al(III) complex system. The facile and spectrum-tunable fluorescent sensor enables rapid detection of PPi in urine and cell lysate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service