Skip to Content
Merck
All Photos(1)

Key Documents

24-5730

Sigma-Aldrich

Potassium thiocyanate solution

0.1 M

Synonym(s):

Potassium rhodanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KSCN
CAS Number:
Molecular Weight:
97.18
Beilstein:
3594799
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
liquid

grade

SAJ special grade

form

liquid

availability

available only in Japan

concentration

0.1 M
1/10 N

pH

5.3-8.7 (25 °C, 97.2 g/L)

density

1 g/mL at 25 °C (lit.)

SMILES string

[K+].[N-]=C=S

InChI

1S/CHNS.K/c2-1-3;/h3H;/q;+1/p-1

InChI key

ZNNZYHKDIALBAK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Botti et al.
The journal of physical chemistry. B, 113(29), 10014-10021 (2009-07-03)
The results of a neutron diffraction experiment performed on aqueous solutions of KSCN are analyzed and discussed in comparison with similar data for KCl and KOH solutions. The effect of the different ions on the structure of water and the
Fei Wang et al.
The Journal of organic chemistry, 77(8), 4148-4151 (2012-03-27)
A concise approach to various benzisothiazol-3(2H)-one derivatives has been developed by copper-catalyzed the reaction of o-bromobenzamide derivatives with potassium thiocyanate (KSCN) in water. The reaction proceeds via a tandem reaction with S-C bond and S-N bond formation.
Altamir Benedito de Sousa et al.
Reproductive toxicology (Elmsford, N.Y.), 23(4), 568-577 (2007-03-06)
A study was performed at term and at weaning to verify the toxic effects of the prenatal exposure to potassium cyanide (KCN) and potassium thiocyanate (KSCN) in pregnant Wistar rats. Females received daily in drinking water the doses: 1, 3
Hongtao Bian et al.
The journal of physical chemistry. B, 116(49), 14426-14432 (2012-11-28)
Microscopic structures and dynamics of aqueous salt solutions were investigated with the ultrafast vibrational energy exchange method and anisotropy measurements. In KSCN aqueous solutions of various concentrations, the rotational time constants of SCN(-) anions are proportional to the viscosities of
Qinjian Zhao et al.
Human vaccines, 2(4), 174-180 (2006-10-03)
The major surface antigen of Hepatitis B virus (HBsAg) is a cysteine-rich, lipid-bound protein with 226 amino acids. Recombinant HBsAg (rHBsAg) with associated lipids can self-assemble into 22-nm immunogenic spherical particles, which are used in licensed Hepatitis B vaccines. Little

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service