Recommended Products
grade
SAJ first grade
Assay
≥99.0%
form
solid
availability
available only in Japan
mp
334 °C (lit.)
SMILES string
[K+].[O-][N+]([O-])=O
InChI
1S/K.NO3/c;2-1(3)4/q+1;-1
InChI key
FGIUAXJPYTZDNR-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 3
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
BMC genomics, 16, 296-296 (2015-04-18)
Propionibacterium freudenreichii (PF) is an actinobacterium used in cheese technology and for its probiotic properties. PF is also extremely adaptable to several ecological niches and can grow on a variety of carbon and nitrogen sources. The aim of this work
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here
Sensors (Basel, Switzerland), 11(4), 4188-4206 (2011-12-14)
Displacement of soil particles caused by erosion influences soil condition and fertility. To date, the cesium 137 isotope ((137)Cs) technique is most commonly used for soil particle tracing. However when large areas are considered, the expensive soil sampling and analysis
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service