Recommended Products
grade
CP
form
liquid
availability
available only in Japan
refractive index
n20/D 1.478 (lit.)
bp
313 °C (lit.)
storage temp.
room temp
SMILES string
O(C(COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)C(=O)CCCCCCC\C=C/CC(O)CCCCCC
InChI key
ZEMPKEQAKRGZGQ-AAKVHIHISA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
557.6 °F
Flash Point(C)
292 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Asian Pacific journal of tropical medicine, 5(8), 630-633 (2012-07-31)
To examine the antidiarrhoeal activity of alcoholic and aqueous seed kernel extract of Mangifera indica (M. indica) on castor oil-induced diarrhoeal activity in Swiss albino mice. Mango seed kernels were processed and extracted using alcohol and water. Antidiarrhoeal activity of
Journal of environmental management, 114, 316-323 (2012-11-23)
Bioenergy production combined with phytoremediation has been suggested to help in solving two critical world problems: the gradual reduction of fossil fuels and soil contamination. The aim of this research was to investigate the potential for the use of Ricinus
Revista brasileira de cirurgia cardiovascular : orgao oficial da Sociedade Brasileira de Cirurgia Cardiovascular, 27(3), 392-400 (2013-01-05)
The aim of the present investigation is to determine if the tissue reaction to the Riccinus communis (mamona) polymer has significant statistical difference compared to the tissue reaction provoked by the titanium implant. Thirty two Cavia porcellus were divided into
Journal of economic entomology, 106(2), 742-746 (2013-06-22)
Leaf-cutting ants are the main herbivores in the New World tropics. Although the toxicity of seed oils against these ants has been poorly investigated, previous results revealed that seed oils exert considerable toxic activity against these insects. This paper analyzes
Parasitology research, 112(2), 611-619 (2012-10-23)
Tick control has been accomplished through the use of synthetic acaricides, which has created resistant individuals, as well as contaminating the environment and nontarget organisms. Substances of plant origin, such as oils and extracts of eucalyptus and neem leaves, have
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service