Skip to Content
Merck
All Photos(1)

Key Documents

15-0160

Sigma-Aldrich

Indigo carmine

JIS special grade, ≥97.5%

Synonym(s):

Acid Blue 74, Indigo-5,5′-disulfonic acid disodium salt, Indigocarmine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H8N2Na2O8S2
CAS Number:
Molecular Weight:
466.35
Colour Index Number:
73015
Beilstein:
4103904
MDL number:
UNSPSC Code:
41116130
PubChem Substance ID:

grade

JIS special grade

Assay

≥97.5%

form

solid

availability

available only in Japan

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc2NC(\C(=O)c2c1)=C3\Nc4ccc(cc4C3=O)S([O-])(=O)=O

InChI

1S/C16H10N2O8S2.2Na/c19-15-9-5-7(27(21,22)23)1-3-11(9)17-13(15)14-16(20)10-6-8(28(24,25)26)2-4-12(10)18-14;;/h1-6,17-18H,(H,21,22,23)(H,24,25,26);;/q;2*+1/p-2/b14-13+;;

InChI key

KHLVKKOJDHCJMG-QDBORUFSSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Montero-Ocampo et al.
Environmental science and pollution research international, 19(9), 3751-3762 (2012-10-12)
In this work, we report in situ studies of UV photoelectrocatalytic discoloration of a dye (indigo carmine) by a TiO(2) thin film in a microreactor to demonstrate the driving force of the applied electrode potential and the dye flow rate
Aurora rings: a novel endoscopic finding to distinguish inverted colonic diverticula from colon polyps.
Michael D Share et al.
Gastrointestinal endoscopy, 77(2), 308-312 (2012-12-05)
Three-dimensional staining of liver segments with an ultrasound contrast agent as an aid to anatomic liver resection.
Junichi Shindoh et al.
Journal of the American College of Surgeons, 215(2), e5-10 (2012-06-12)
Paola Iovino et al.
Gastrointestinal endoscopy, 77(2), 233-240 (2012-12-04)
Chromo-zoom endoscopy has been demonstrated to be valuable in assessing the degree of intestinal villous atrophy in patients with suspected celiac disease. To evaluate the diagnostic accuracy of chromo-zoom endoscopy in patients with difficult diagnosis because of nonconcordant test results
Eleanor M Pritchard et al.
Macromolecular bioscience, 13(3), 311-320 (2013-01-26)
Sericin removal from the core fibroin protein of silkworm silk is a critical first step in the use of silk for biomaterial-related applications, but degumming can affect silk biomaterial properties, including molecular weight, viscosity, diffusivity and degradation behavior. Increasing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service