Skip to Content
Merck
All Photos(1)

Documents

01-5300

Sigma-Aldrich

1-Pentanol

SAJ first grade, ≥97.0%

Synonym(s):

n-Amyl alcohol, Pentyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4OH
CAS Number:
Molecular Weight:
88.15
Beilstein:
1730975
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

grade

SAJ first grade

vapor density

3 (vs air)

vapor pressure

1 mmHg ( 13.6 °C)
1.5 mmHg ( 20 °C)

Assay

≥97.0%

form

liquid

expl. lim.

10 %, 100 °F

availability

available only in Japan

refractive index

n20/D 1.409 (lit.)

pH

7

bp

136-138 °C (lit.)

mp

−78 °C (lit.)

density

0.811 g/mL at 25 °C (lit.)

SMILES string

CCCCCO

InChI

1S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3

InChI key

AMQJEAYHLZJPGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Jae Jeon et al.
Journal of biotechnology, 135(2), 127-133 (2008-04-26)
A thermostable alcohol dehydrogenase (ADH-I) isolated from the potential thermophilic ethanologen Geobacillus thermoglucosidasius strain M10EXG has been characterised. Inverse PCR showed that the gene (adhI) was localised with 3-hexulose-6-phosphate synthase (HPS) and 6-phospho-3 hexuloisomerase (PHI) on its genome. The deduced
Sebastian Höfener et al.
Physical chemistry chemical physics : PCCP, 10(23), 3390-3399 (2008-06-07)
In the recent years, Slater-type geminals (STGs) have been used with great success to expand the first-order wave function in an explicitly-correlated perturbation theory. The present work reports on this theory's implementation in the framework of the Turbomole suite of
Anna L Barnette et al.
Langmuir : the ACS journal of surfaces and colloids, 26(21), 16299-16304 (2010-08-26)
The origin and role of tribochemical reaction products formed while sliding silicon oxide surfaces in the presence of adsorbed alcohol molecules in equilibrium with the vapor phase were studied. Wear and friction coefficient studies with varying contact loads and n-pentanol
Gizelle Cristina Bedendo et al.
Journal of chromatography. A, 1217(42), 6449-6454 (2010-09-14)
A sensitive and precise analysis using hollow fiber renewal liquid membrane (HFRLM) extraction followed by high performance liquid chromatography-tandem mass spectrometry (LC-MS/MS) is described for determination of five sulfonamides in honey samples. In this procedure, the organic solvent introduced directly
Nadav Amdursky et al.
The journal of physical chemistry. A, 115(24), 6481-6487 (2011-05-19)
Time-resolved emission techniques were employed to study the nonradiative process of thioflavin-T (ThT) in 1-propanol, 1-butanol, and 1-pentanol as a function of the hydrostatic pressure. Elevated hydrostatic pressure increases the alcohol viscosity, which in turn strongly influences the nonradiative rate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service