Skip to Content
Merck
All Photos(1)

Key Documents

01-4780

Sigma-Aldrich

Ammonium metavanadate

JIS special grade, ≥99.0%

Synonym(s):

Ammonium vanadate(V)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4VO3
CAS Number:
Molecular Weight:
116.98
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

JIS special grade

Assay

≥99.0%

form

solid

reaction suitability

reagent type: catalyst
core: vanadium

availability

available only in Japan

density

2.32 g/cm3 at 25 °C (lit.)

SMILES string

N.O[V](=O)=O

InChI

1S/H3N.H2O.2O.V/h1H3;1H2;;;/q;;;;+1/p-1

InChI key

YBVKNHXQSRDWAA-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - STOT RE 1 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tridib Chakraborty et al.
Biochimica et biophysica acta, 1772(1), 48-59 (2006-12-19)
Carcinogen-induced formation of DNA adducts and other types of DNA lesions are the critical molecular events in the initiation of chemical carcinogenesis and modulation of such events by chemopreventive agents could be an important step in limiting neoplastic transformation in
H K Parsadanian et al.
Neurotoxicology, 19(4-5), 561-564 (1998-09-24)
Vanadium and its derivatives are the well-known environmental pollutants. We obtained that ammonium vanadate suppressed the alkaline (AlP) and acid (AcP) phosphatases activity in the variety of tissues. Vanadate inhibited the enzymes that take part in phosphoryl transfer reactions. Successive
Kazuo Kobayashi et al.
Toxicology, 228(2-3), 162-170 (2006-09-22)
Metallothionein (MT) is a low-molecular-weight cysteine-rich protein which has a high affinity for metals. The synthesis of MT is induced by heavy metals such as cadmium and zinc. However, little is known about the induction of MT by tetravalent or
Jean P Gaffney et al.
Biochemistry, 48(49), 11609-11611 (2009-11-20)
Some ascidians sequester vanadium and other metal ions that are bound and transported in higher organisms by transferrin. The ascidian Ciona intestinalis has a monolobal transferrin (nicatransferrin) in its plasma. The binding of vanadium(V) to nicatransferrin was investigated by using
Ghada Abdelhamid et al.
Toxicology in vitro : an international journal published in association with BIBRA, 24(6), 1554-1561 (2010-07-06)
Recent studies demonstrated the carcinogenicity and the mutagenicity of vanadium compounds. In addition, vanadium (V(5+)) was found to enhance the effects of other genotoxic agents. However, the mechanism by which V(5+) induce toxicity remain unknown. In the current study we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service