Skip to Content
Merck
All Photos(1)

Key Documents

PX1467

Supelco

Potassium Fluoride

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Potassium Fluoride, potassium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FK
CAS Number:
Molecular Weight:
58.10
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-151-5
form:
solid

Agency

meets USP/NF

Quality Level

vapor pressure

1.3 hPa ( 885 °C)

Assay

≥99.0% (precipitation titration)

form

solid

impurities

≤0.01 meq/g Titrable base
≤0.03 meq/g Titrable acid
≤0.1% Potassium fluosilicate (K2SiF6)

pH

8-9 (20 °C, 50 g/L in H2O)

bp

1505 °C/1013 hPa

mp

846 °C

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.005%

cation traces

Fe: ≤0.001%
Na: ≤0.2%
heavy metals (as Pb): ≤0.001%

shipped in

ambient

storage temp.

room temp

SMILES string

[K+].[F-]

InChI

1S/FH.K/h1H;/q;+1/p-1

InChI key

NROKBHXJSPEDAR-UHFFFAOYSA-M

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Analysis Note

Assay: 99.0% min
Chloride (Cl): 0.005% max
Heavy metals (as Pb): 0.001% max
Iron (Fe): 0.001% max
Potassium fluosilicate (K2SiF6): 0.1% max
Sodium (Na): 0.2% max
Sulfate (SO4): 0.005% max
Titrable acid: 0.03meq/g max
Titrable base: 0.01meq/g max

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service