Skip to Content
Merck
All Photos(2)

Key Documents

MABT529

Sigma-Aldrich

Anti-Integrin beta 1 (CD29) Antibody

mouse monoclonal, 13C4.1

Synonym(s):

Integrin beta-1, CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Integrin beta-1 (CD29) Antibody, clone 13C4.1, clone 13C4.1, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

13C4.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1λ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITGB1(3688)

General description

Integrin beta-1 (UniProt P05556; also known as CD29, Glycoprotein Iia, GPIIA, Integrin VLA-4 subunit beta, Very late activation protein, beta polypeptide) is encoded by the ITGB1 (also known as FNRB, MDF2, MSK12, VLA-BETA, VLAB) gene (Gene ID 3688) in human. Integrin beta-1 is a 130 kDa transmembrane glycoprotein that interact with various integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptor complexes that bind to specific extracellular matrix proteins. Integrin receptors regulate a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor growth and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to the activation of signal transduction pathways and regulation of gene expression.

Specificity

Clone 13C4.1 targets an epitope within the membrane-proximal extracellular region present in all five spliced isoforms of human Integrin beta-1 reported by UniPort (P05556).

Immunogen

GST-tagged recombinant human Integrin beta-1 fragment corresponding to the membrane-proximal extracellular region.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected Integrin beta-1 in human breast cancer, cerebral cortex, and lung tissue sections.
Research Category
Cell Structure
This mouse monoclonal Anti-Integrin beta-1 (CD29), clone 13C4.1, Cat. No. MABT529, is validated for use in Immunohistochemistry (Paraffin) and Western Blotting for the detection of Integrin beta-1.

Quality

Evaluated by Western Blotting in U937 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Integrin beta-1 in 10 µg of U937 cell lysate.

Target description

~150 kDa observed. Target band size appears larger than the calculated molecular weights of 88.42/87.45/91.62/91.03/88.88 kDa (isoform Beta-1A/Beta-1B/Beta-1C/Beta-1C-2/Beta-1D pro-form) and 86.19/85.22/89.40/88.81/86.66 kDa (mature isoform Beta-1A/Beta-1B/Beta-1C/Beta-1C-2/Beta-1D) due to glycosylation. Uncharacterized bands may be observed in some lysate(s).

Linkage

Replaces: 04-1109

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trang Thi-Huynh Le et al.
American journal of cancer research, 12(1), 176-197 (2022-02-11)
Metastatic and castration-resistant disease is a fatal manifestation of prostate cancer (PCa). The mechanism through which resistance to androgen deprivation in PCa is developed remains largely unknown. Our understanding of the tumor microenvironment (TME) and key signaling pathways between tumors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service