Skip to Content
Merck
All Photos(1)

Key Documents

MABE1067

Sigma-Aldrich

Anti-ESET (SETDB1) Antibody, clone F0808

clone F0808, from mouse

Synonym(s):

Histone-lysine N-methyltransferase SETDB1, EC: 2.1.1.43, ERG-associated protein with SET domain, SET domain bifurcated 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

F0808, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Setdb1(84505)

General description

Histone-lysine N-methyltransferase SETDB1 (UniProt: O88974; also known as EC: 2.1.1.43, ERG-associated protein with SET domain, ESET, SET domain bifurcated 1, SETDB1) is encoded by the Setdb1 (also known as Eset, Kiaa0067) gene in murine species. SETDB1 is a ubiquitously expressed histone methyltransferase with multiple zinc-binding sites that specifically trimethylates lysine 9 of histone H3 immediately downstream of transcription start sites marked with H3K4me3 to establish the bivalent domain. It is targeted to histone H3 by TRIM28/TIF1B, a factor recruited by Krüppel-associated box (KRAB) zinc-finger proteins. The H3K4/H3K9me3 signature keeps RNA polymerase II (Pol II) paused at the proximal promoter and maintains gene expression at low levels, but in a poised state that is ready for activation in lineage-committed cells. It functions mainly in euchromatin regions, thereby playing a key role in the silencing of euchromatic genes. It probably acts by forming a complex with MBD1 and ATF7IP that represses transcription and couples DNA methylation and histone lysine 9 trimethylation. SETDB1 is reported to inhibit adipogenesis and its depletion induces preadipocyte differentiation and adipogenesis in mesenchymal stem cells and preadipocytes. Expression of Cebpa and Pparg genes that are its key targets of SETDB1 is enhanced when SETDB1 is knocked down in undifferentiated cells. Murine SETDB1 contains two Tudor domains (aa 257-320 and 347-403) and pre-SET (aa 744-817), SET (aa 820-1282), and post-SET (aa 1291-1307) domains that are required for its methyltransferase activity. (Ref.: Matsumura, Y., et al. (2015). Mol. Cell 60(4); 584-596).

Specificity

Clone F0808 specifically detects Histone-lysine N-methyltransferase SETDB1. It targets an epitope with in the internal region.

Immunogen

His-tagged recombinant fragment corresponding to 213 amino acids from the internal region of murine SETDB1.

Application

Anti-ESET (SETDB1), clone F0808, Cat. No. MABE1067, is a highly specific mouse monoclonal antibody that targets Histone-lysine N-methyltransferase SETDB1 and has been tested for use in Chromatin Immunoprecipitation (ChIP), Immunoprecipitation, and Western Blotting.
Chromatin Immunoprecipitation Analysis (ChIP): A representative lot detected ESET (SETDB1) in Chromatin Immunoprecipitation applications (Matsumura, Y., et. al. (2015). Mol Cell. 60(4):584-96).

Immunoprecipitation Analysis: A representative lot immunoprecipitated ESET (SETDB1) in Immunoprecipitation applications (Matsumura, Y., et. al. (2015). Mol Cell. 60(4):584-96).

Quality

Evaluated by Western Blotting in 129/S6/SvEv mouse embryonic stem cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected ESET (SETDB1) in 129/S6/SvEv mouse embryonic stem cell lysate.

Target description

~180/200 kDa observed; 144.55 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service