Skip to Content
Merck
All Photos(1)

Key Documents

MAB3568

Sigma-Aldrich

Anti-Myosin Antibody, smooth muscle heavy chain, SM1 & SM2, clone ID8

clone ID8 (SMMS-1), Chemicon®, from mouse

Synonym(s):

Anti-CCDC128, Anti-KLRAQ1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ID8 (SMMS-1), monoclonal

species reactivity

human, bovine, pig, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYH11(4629)

Application

This Anti-Myosin Antibody, smooth muscle heavy chain, SM1 & SM2, clone ID8 is validated for use in IP, WB, IC, IH(P) for the detection of Myosin.
Western blot (see application notes below). Suggested blocking buffer is TBS-Tween with 2% BSA. Suggested dilution buffer is TBS-Tween with 0.05% sodium azide. Preferred gel percentage is 5% (see application notes).

Immunohistochemistry on frozen and paraffin embedded tissue sections. Suggested fixation for frozen tissue sections is acetone fix for 6 minutes at room temperature. For formalin fixed paraffin embedded tissue sections: microwave in 0.01M citrate buffer (pH 6.0) for 8-10 minutes (note that all microwaves differ and adjustments may need to be made) follow with enzyme digestion (0.01% pronase for 10 mintues). Suggested blocking agent is fetal bovine serum.

Immunocytochemistry

Immunoprecipitation. Suggested extraction buffer is 20 mM Tris-HCl, pH 7.4, 150 mM NaCl, 1% Triton X-100, 0.1% SDS, 0.5% deoxycholic acid-NaCl and 0.5 mM PMSF. Final reaction volume is 1 mL and suggested capture agent is agarose conjugated anti-mouse IgG.

Optimal working dilutions must be determined by the end user.

APPLICATION NOTES FOR MAB3568

WESTERN BLOT

To achieve good resolution of myosin heavy chain isoforms with distinct molecular weight (200 - 2004 kDa), the following procedure should be followed: 1). Pyrophoshate extraction buffer for sample preparation (see below); run SDS-PAGE in 5% gel. Important: for better resolution of the MHC bands, use electrophoretic buffer with pH 8.2 (i.e. 0.1 less than standard), and prepare resolving gel (5%) with pH 9.0 (not 8.8 as usual). Also help thorough degasing of the resolving gel mixture (H2O, acrylamide, EDTA, pH 9.0, before (!) adding SDS, TEMED and APS). Run SDS-PAGE longer than after the dye front runs off (use 200 kDa MW markers and let it′s 200 kDa band run at least to the middle of 8X8 gel (using a big size gel (not the mini-gel!) will enhance the quality of MHC band resolution).

Pyrophosphate extraction buffer: (40mM Na4P2O7x10H2O, 1mM MgCl2, 1mM EGTA (add KOH to dissolve EGTA), PMSF, pH 9.5). To extract acto-myosin from tissues/cells, shake minced tissue or cells in cold extraction buffer 1 hr on ice bath (0oC), centrifuge @10,000g for 10 min at +2-8 oC, take supernatant and mix it 1:1 with standard Laemmli sample buffer, boil, run SDS-PAGE in 5% gel (see above).

Physical form

Format: Purified

Analysis Note

Control
POSITIVE CONTROL: Smooth muscle (e.g. aterial tunica media). Negative control: any nonmuscle tissue (e.g. arterial tunica adentitial).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deli Liu et al.
Cancer research, 74(18), 5045-5056 (2014-08-02)
Spontaneously occurring canine mammary cancer represents an excellent model of human breast cancer, but is greatly understudied. To better use this valuable resource, we performed whole-genome sequencing, whole-exome sequencing, RNA-seq, and/or high-density arrays on twelve canine mammary cancer cases, including
Pei F Lai et al.
The Journal of physiology, 594(21), 6369-6393 (2016-11-02)
Over 15 million babies are born prematurely each year with approximately 1 million of these babies dying as a direct result of preterm delivery. β2 -Adrenoreceptor agonists that act via cAMP can reduce uterine contractions to delay preterm labour, but
Brandan Walters et al.
Cells, 10(11) (2021-11-28)
Adipose-derived stem cells (ASCs) are an abundant and easily accessible multipotent stem cell source with potential application in smooth muscle regeneration strategies. In 3D collagen hydrogels, we investigated whether sustained release of growth factors (GF) PDGF-AB and TGF-β1 from GF-loaded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service