Skip to Content
Merck
All Photos(1)

Key Documents

DX2090

Supelco

p-Dioxane

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

p-Dioxane, 1,4-Dioxane, 1

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
-O(CH2)2O(CH2)2-
CAS Number:
MDL number:
UNSPSC Code:
12191504
EC Index Number:
204-661-8
grade:
reagent
bp:
101.5 °C/1013 hPa
vapor pressure:
41 hPa ( 20 °C)

grade

reagent

Quality Level

Agency

meets USP/NF

vapor pressure

41 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

shelf life

60 mo. (from manufacture date)

dilution

(for analytical testing)

impurities

≤0.0016 meq/g Titrable acid
≤0.005% Peroxide (as H2O2)
≤0.01% Carbonyl (as HCHO)
≤0.05% Water (H2O)

evapn. residue

≤0.005%

color

APHA: ≤20

pH

6-8 (20 °C, 500 g/L in H2O)

bp

101.5 °C/1013 hPa

mp

12 °C

transition temp

flash point 11 °C

density

1.03 g/cm3 at 20 °C

shipped in

ambient

storage temp.

no temp limit

SMILES string

O1CCOCC1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Expiration dateExp 60 months from mfg dateAssay (GC)99.0% minBHT stabilizer presentConformsCarbonyl (as HCHO)0.01% maxColor (APHA)20 maxEvaporation residue0.005% maxFreezing point11.0°C minIdentity (IR-spectrum)ConformsPeroxide (as H2O2)0.005% maxTitrable acid0.0016meq/g maxWater (H2O)0.05% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service