Skip to Content
Merck
All Photos(1)

Key Documents

CX0914

Supelco

1-Chlorobutane

OmniSolv®

Synonym(s):

1-Chlorobutane, Butyl Chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3Cl
CAS Number:
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-696-6
technique(s):
HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable
bp:
79 °C/1013 hPa
vapor pressure:
110 hPa ( 20 °C)

vapor pressure

110 hPa ( 20 °C)

Quality Level

product line

OmniSolv®

Assay

≥99.5% (GC)

form

liquid

technique(s)

HPLC: suitable
LC/MS: suitable
gas chromatography (GC): suitable

impurities

≤0.01% Water (H2O)

evapn. residue

≤1 ppm

color

APHA: ≤10

bp

79 °C/1013 hPa

mp

-123 °C

transition temp

flash point -17 °C

solubility

0.5 g/L

density

0.886 g/cm3 at 20 °C

UV cutoff

≤220 nm

UV absorption

λ: 220 nm Amax: ≤1.00
λ: 225 nm Amax: ≤0.50
λ: 233 nm Amax: ≤0.10
λ: 240 nm Amax: ≤0.05
λ: 270 nm Amax: ≤0.01
λ: 300 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

SMILES string

ClCCCC

InChI

1S/C4H9Cl/c1-2-3-4-5/h2-4H2,1H3

InChI key

VFWCMGCRMGJXDK-UHFFFAOYSA-N

Application

OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.5% minColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)1.0ppb maxFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 220 nm1.00AU maxUV Abs. at 225 nm0.50AU maxUV Abs. at 233 nm0.10AU maxUV Abs. at 240 nm0.05AU maxUV Abs. at 270 nm0.01AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off220nm maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service